ENA308583643 Enamine

1-(1,3-thiazol-2-yl)piperidine-3-carboxylic acid

  • MDL number MFCD09054880
  • Cas Number : 927803-56-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68166
Availability details for
Quantity
  • Availability
  • Add to Cart