ENA308583651 Enamine

1-cyclohexylbutan-1-amine

  • MDL number MFCD12154077
  • Cas Number : 854919-17-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68226

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68226
Availability details for
Quantity
  • Availability
  • Add to Cart