ENA308583653 Enamine

4-(chloromethyl)-2-(2-fluorophenyl)-1,3-thiazole

  • MDL number MFCD09729635
  • Cas Number : 138128-84-4
  • Molecular Weight : 227.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68238
Availability details for
Quantity
  • Availability
  • Add to Cart