ENA308583655 Enamine

3-(4-bromo-1H-pyrazol-1-yl)-N,N-dimethylpropanamide

  • MDL number MFCD12824378
  • Cas Number : 1184274-21-2
  • Molecular Weight : 246.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68257

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68257
Availability details for
Quantity
  • Availability
  • Add to Cart