ENA308583656 Enamine

2-pentyl-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD11198617
  • Cas Number : 69588-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68258

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68258
Availability details for
Quantity
  • Availability
  • Add to Cart