ENA308583657 Enamine

2-(diethylamino)-2-phenylacetamide

  • MDL number MFCD17167148
  • Cas Number : 732938-75-9
  • Molecular Weight : 206.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(diethylamino)-2-phenylacetamide
Inchi Code 1S/C12H18N2O/c1-3-14(4-2)11(12(13)15)10-8-6-5-7-9-10/h5-9,11H,3-4H2,1-2H3,(H2,13,15)
Inchi Key GCJFHFOHKMTRKU-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68268
Availability details for
Quantity
  • Availability
  • Add to Cart