ENA308583658 Enamine

2-(5-chlorothiophen-2-yl)-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD11186361
  • Cas Number : 1218116-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68282

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P312,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-68282
pictograms GHS07
hstatements H302,H313,H315,H319,H333,H335
Availability details for
Quantity
  • Availability
  • Add to Cart