ENA308583661 Enamine

4-bromo-1-(prop-2-en-1-yl)-1H-pyrazole

  • MDL number MFCD12780035
  • Cas Number : 13369-72-7
  • Molecular Weight : 187.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68292
Availability details for
Quantity
  • Availability
  • Add to Cart