ENA308583673 Enamine

2-(2-methyl-2,3-dihydro-1H-indole-1-carbonyl)cyclohexane-1-carboxylic acid

  • MDL number MFCD06059147
  • Cas Number : 1052596-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68319

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68319
Availability details for
Quantity
  • Availability
  • Add to Cart