ENA308583674 Enamine

5-(2,4,6-trimethylphenyl)-2,3-dihydro-1,3,4-oxadiazole-2-thione

  • MDL number MFCD11201465
  • Cas Number : 168638-04-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68321

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68321
Availability details for
Quantity
  • Availability
  • Add to Cart