ENA308583676 Enamine

6-bromo-8-methyl-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09740135
  • Cas Number : 954563-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68325
Availability details for
Quantity
  • Availability
  • Add to Cart