ENA308583689 Enamine

1-(3-chlorophenyl)-4-methylpentan-2-one

  • MDL number MFCD11936553
  • Cas Number : 1175909-18-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68346

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68346
Availability details for
Quantity
  • Availability
  • Add to Cart