ENA308583690 Enamine

3-ethynyl-N-[(thiophen-2-yl)methyl]aniline

  • MDL number MFCD11138203
  • Cas Number : 1019586-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68347

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68347
Availability details for
Quantity
  • Availability
  • Add to Cart