ENA308583693 Enamine

(4-aminooxan-4-yl)methanol

  • MDL number MFCD04115578
  • Cas Number : 720706-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68350

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-68350
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart