ENA308583694 Enamine

3-(3-aminopropyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11131200
  • Cas Number : 1019391-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68351

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68351
Availability details for
Quantity
  • Availability
  • Add to Cart