ENA308583697 Enamine

3-(3-nitrophenyl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00452056
  • Cas Number : 59301-20-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68354

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68354
Availability details for
Quantity
  • Availability
  • Add to Cart