ENA308583698 Enamine

ethyl 2-(3-methyl-1,2,4-oxadiazol-5-yl)acetate

  • MDL number MFCD13705352
  • Cas Number : 1216294-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68355

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68355
Availability details for
Quantity
  • Availability
  • Add to Cart