ENA308583702 Enamine

1-(4-hydroxyphenyl)-1-methylurea

  • MDL number MFCD11126508
  • Cas Number : 106837-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68367
Availability details for
Quantity
  • Availability
  • Add to Cart