ENA308583709 Enamine

2-(4-bromophenyl)-4,5,6,7-tetrahydro-1,3-benzothiazol-7-one

  • MDL number MFCD17167162
  • Cas Number : 1258649-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68375
Availability details for
Quantity
  • Availability
  • Add to Cart