ENA308583712 Enamine

1-formamidocyclopentane-1-carboxylic acid

  • MDL number MFCD14589581
  • Cas Number : 15026-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68379

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68379
Availability details for
Quantity
  • Availability
  • Add to Cart