ENA308583713 Enamine

[1-(methylamino)cyclopentyl]methanol

  • MDL number MFCD12781674
  • Cas Number : 1094072-11-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67...69
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68380

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-68380
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart