ENA308583716 Enamine

4-amino-N-methoxy-N-methylbenzamide

  • MDL number MFCD12095384
  • Cas Number : 186252-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68384

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68384
Availability details for
Quantity
  • Availability
  • Add to Cart