ENA308583721 Enamine

4-amino-2-methylbenzene-1-sulfonamide

  • MDL number MFCD11621528
  • Cas Number : 94109-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68390

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68390
Availability details for
Quantity
  • Availability
  • Add to Cart