ENA308583727 Enamine

2-[(1-phenyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]-1-(1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD06388969
  • Cas Number : 801235-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68397

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68397
Availability details for
Quantity
  • Availability
  • Add to Cart