ENA308583755 Enamine

methyl 2-(bromomethyl)-5-fluorobenzoate

  • MDL number MFCD04114323
  • Cas Number : 138786-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41...43
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68441
Availability details for
Quantity
  • Availability
  • Add to Cart