ENA308583763 Enamine

2-[(2-methylphenyl)amino]-1-(piperidin-1-yl)ethan-1-one

  • MDL number MFCD01700406
  • Cas Number : 93865-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68451
Availability details for
Quantity
  • Availability
  • Add to Cart