ENA308583765 Enamine

4-(1H-imidazol-1-yl)-3-methylaniline

  • MDL number MFCD11106415
  • Cas Number : 461664-00-6
  • Molecular Weight : 173.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68453

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-68453
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart