ENA308583768 Enamine

3-ethynyl-N-[(furan-2-yl)methyl]aniline

  • MDL number MFCD11138205
  • Cas Number : 369363-98-4
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68457

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68457
Availability details for
Quantity
  • Availability
  • Add to Cart