ENA308583777 Enamine

ethyl 4-bromo-3-(chlorosulfonyl)benzoate

  • MDL number MFCD12169502
  • Cas Number : 1155911-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68471
Availability details for
Quantity
  • Availability
  • Add to Cart