ENA308583778 Enamine

1-(3-phenylpropyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD12868674
  • Cas Number : 1178833-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68472

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68472
Availability details for
Quantity
  • Availability
  • Add to Cart