ENA308583779 Enamine

5-(2-methoxyphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD01710631
  • Cas Number : 5711-59-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68473

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68473
Availability details for
Quantity
  • Availability
  • Add to Cart