ENA308583785 Enamine

7-nitro-2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride

  • MDL number MFCD09811971
  • Cas Number : 5279-54-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68481
Availability details for
Quantity
  • Availability
  • Add to Cart