ENA308583791 Enamine

3-(2-oxo-1,3-oxazolidin-3-yl)-3-phenylpropanoic acid

  • MDL number MFCD11215176
  • Cas Number : 150812-64-9
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68493

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68493
Availability details for
Quantity
  • Availability
  • Add to Cart