ENA308583793 Enamine

methyl 2-(4-oxo-4,5-dihydro-1H-1,5-benzodiazepin-2-yl)acetate

  • MDL number MFCD05854735
  • Cas Number : 91392-96-0
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68495
Availability details for
Quantity
  • Availability
  • Add to Cart