ENA308583795 Enamine

2-methyl-3H,4H-pyrido[2,3-b]pyrazin-3-one

  • MDL number MFCD11107149
  • Cas Number : 128102-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68497

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68497
Availability details for
Quantity
  • Availability
  • Add to Cart