ENA308583800 Enamine

2-benzylcyclohexan-1-amine

  • MDL number MFCD09880441
  • Cas Number : 72436-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68503

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68503
Availability details for
Quantity
  • Availability
  • Add to Cart