ENA308583805 Enamine

3-(4-tert-butylphenyl)-1,2-oxazol-5-amine

  • MDL number MFCD07382880
  • Cas Number : 794566-80-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68546

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68546
Availability details for
Quantity
  • Availability
  • Add to Cart