ENA308583813 Enamine

3-bromo-2-hydroxy-5-methoxybenzaldehyde

  • MDL number MFCD00091938
  • Cas Number : 50343-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68557

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68557
Availability details for
Quantity
  • Availability
  • Add to Cart