ENA308583827 Enamine

tert-butyl N-[2-(3-aminophenoxy)ethyl]carbamate

  • MDL number MFCD11922407
  • Cas Number : 246240-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68575
Availability details for
Quantity
  • Availability
  • Add to Cart