ENA308583829 Enamine

4-(pyridin-2-yl)piperazine-1-carbothioamide

  • MDL number MFCD09803957
  • Cas Number : 875229-19-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68578

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68578
Availability details for
Quantity
  • Availability
  • Add to Cart