ENA308583830 Enamine

1-butyl-5-(5-fluoro-2-hydroxybenzoyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD10649405
  • Cas Number : 1087634-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-68579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68579
Availability details for
Quantity
  • Availability
  • Add to Cart