ENA308583837 Enamine

2-[(3-methylphenyl)amino]-1-(piperidin-1-yl)ethan-1-one

  • MDL number MFCD11149895
  • Cas Number : 93865-38-4
  • Molecular Weight : 232.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-68592

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68592
Availability details for
Quantity
  • Availability
  • Add to Cart