ENA308583841 Enamine

2-(5-chlorothiophen-2-yl)acetonitrile

  • MDL number MFCD09817278
  • Cas Number : 42520-90-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68600
Availability details for
Quantity
  • Availability
  • Add to Cart