ENA308583845 Enamine

4-chloro-6-ethyl-2-(pyridin-4-yl)pyrimidine

  • MDL number MFCD12137658
  • Cas Number : 1153412-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68621

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68621
Availability details for
Quantity
  • Availability
  • Add to Cart