ENA308583846 Enamine

1-(2,6-dichlorophenyl)-5-methyl-1H-1,2,4-triazole-3-carboxylic acid

  • MDL number MFCD12106486
  • Cas Number : 1154101-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68622

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68622
Availability details for
Quantity
  • Availability
  • Add to Cart