ENA308583847 Enamine

3-[(2-aminophenyl)sulfanyl]propanamide

  • MDL number MFCD11206112
  • Cas Number : 98898-02-3
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68624
Availability details for
Quantity
  • Availability
  • Add to Cart