ENA308583849 Enamine

4-fluoro-2-nitro-N-(propan-2-yl)aniline

  • MDL number MFCD00798379
  • Cas Number : 146366-00-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68627

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68627
Availability details for
Quantity
  • Availability
  • Add to Cart