ENA308583854 Enamine

3-ethynyl-N-[(pyridin-4-yl)methyl]aniline

  • MDL number MFCD11138208
  • Cas Number : 1019511-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68632
Availability details for
Quantity
  • Availability
  • Add to Cart