ENA308583856 Enamine

(3-aminopropyl)(3-phenylpropyl)amine

  • MDL number MFCD12803558
  • Cas Number : 67884-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68634

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68634
Availability details for
Quantity
  • Availability
  • Add to Cart