ENA308583870 Enamine

2-amino-2-[3-(trifluoromethyl)phenyl]propan-1-ol

  • MDL number MFCD12781980
  • Cas Number : 1183323-24-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68653

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68653
Availability details for
Quantity
  • Availability
  • Add to Cart